missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(-)-Norgestrel, ≥99%, MilliporeSigma™ Supelco™
D(â)-Norgestrel is a hormonal medication and a contraceptive used by women.
Supplier: Merck Emd Millipore N22601G
Specifications
| Poly Bottle | |
| Neat | |
| MFCD00199013 | |
| Analytical Standard | |
| 312.45 | |
| Analytical Standard |
| 797-63-7 | |
| C21H28O2 | |
| 13 beta-Ethyl-17 alpha-ethynyl-17 beta-hydroxygon-4-en-3-one; 17 alpha-Ethynyl-18-homo-19-nor-testosterone; 18,19-Dinor-13 beta-ethyl-17 beta-hydroxy-4-pregnen-20-yn-3-one; 18,19-Dinor-4-pregnen-20-yn-3-one; Levonorgestrel | |
| 1 g | |
| ≥99% |