missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Methionine 99.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 149.21 |
| Percent Purity | ≥99.0% (T) |
| Grade | Laboratory |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | D-Methionine |
Safety and Handling
| EINECSNumber | (2)-1254 |
| RTECSNumber | PD0455000 |
| TSCA | Yes |