missing translation for 'onlineSavingsMsg'
Learn More

D-Mannitol 99.0+%, TCI America™

Catalog No. M0044500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0044500G 500 g
M004425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0044500G Supplier TCI America Supplier No. M0044500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69-65-8 | C6H14O6 | 182.17 g/mol

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Melting Point 168°C
Color White
Quantity 500 g
Formula Weight 182.17
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Mannitol
EINECSNumber (8)-0049&(9)-1375
RTECSNumber OP2060000
TSCA Yes