missing translation for 'onlineSavingsMsg'
Learn More

D-Mannitol, ACS reagent

Catalog No. AC423922500
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423922500 250 g Plastic bottle
AC423920010 1 kg Plastic bottle
AC423920050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC423922500 Supplier Thermo Scientific Chemicals Supplier No. 42392 2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69-65-8 | C6H14O6 | 182.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Melting Point 165.0°C to 170.0°C
Color White
Ignition Residue 0.01% max.
Loss on Drying 0.05% max. (5 g, 105°C, 2 hrs)
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 250 g
Beilstein 01, IV, 2841
Merck Index 15, 5810
Specific Rotation +23.3° to +24.3° (25°C, 589nm) (Reagent ACS-directives)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in pyridine and aniline,slightly soluble in lower alcohols,practically insoluble in most other common,organic solvents
Formula Weight 182.17
Grade ACS Reagent
Insoluble Matter 0.01% max
Titratable Acid 0.0008 meq/g max
Physical Form Crystalline Powder
Chemical Name or Material D-Mannitol
Show More Show Less
EINECSNumber 200-711-8

RUO – Research Use Only