missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Mannitol, 98+%

CAS: 69-65-8 | C6H14O6 | 182.17 g/mol

Supplier:  Thermo Scientific Chemicals 125340050

Quantity 5 kg
Packaging Plastic drum

 Afficher plus de versions de ce produit

Numéro de catalogue. AC125340050


Il en reste null

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

69-65-8
182.17
FBPFZTCFMRRESA-UHFFFAOYNA-N
6251
OCC(O)C(O)C(O)C(O)CO
C6H14O6
MFCD00064287
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
CHEBI:16899

Spécifications

D-Mannitol
165.0°C to 170.0°C
0.3% max. (105°C, 4 hrs)
98% min. (ELSD) (HPLC)
C6H14O6
MFCD00064287
15, 5810
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
FBPFZTCFMRRESA-UHFFFAOYNA-N
(2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
6251
182.17
Crystalline Powder
69-65-8
White
Authentic
Plastic drum
5 kg
01, IV, 2841
+23° to +25° (20°C, 589nm) (PHEUR)
Solubility in water: soluble. Other solubilities: soluble in pyridine and aniline,slightly soluble in lower alcohols,practically insoluble in most other common,organic solvents
OCC(O)C(O)C(O)C(O)CO
182.17
CHEBI:16899
98+%
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

EINECSNumber : 200-711-8

FDS
Documents

Documents

Promotions

Promotions