missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Mannitol, 97+%

CAS: 69-65-8 | C6H14O6 | 182.17 g/mol

$70.86 - $414.34

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
IUPAC Name (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1403030
View Documents Promotion Details
Thermo Scientific Chemicals
A1403030
250 g
Each for $70.86
Only null left
 
AAA140300B
View Documents Promotion Details
Thermo Scientific Chemicals
A140300B
1000 g
Each for $134.75
Only null left
 
AAA140300I
View Documents Promotion Details
Thermo Scientific Chemicals
A140300I
5000 g
Each for $414.34
Only null left
 
Description

Description

D-Mannitol is used in the determination of glomerular filtration rate. It is used as an inert osmotic control substance. Further, it is used in the treatment of oliguric renal failure. It is also used as a sweetener for people with diabetes, in chewing gums and acts as an adulterant for heroin, methamphetamines and cocaine. It finds application as a research tool in cell biological studies, usually to control osmolarity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Mannitol is used in the determination of glomerular filtration rate. It is used as an inert osmotic control substance. Further, it is used in the treatment of oliguric renal failure. It is also used as a sweetener for people with diabetes, in chewing gums and acts as an adulterant for heroin, methamphetamines and cocaine. It finds application as a research tool in cell biological studies, usually to control osmolarity.

Solubility
Soluble in water, alcohol, aniline, pyridine and glycerol. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

69-65-8
182.17
FBPFZTCFMRRESA-UHFFFAOYNA-N
6251
(2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
C6H14O6
MFCD00064287
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
CHEBI:16899
OCC(O)C(O)C(O)C(O)CO

Specifications

69-65-8
1.489
Odorless
MFCD00064287
14,5745
Soluble in water,alcohol,aniline,pyridine and glycerol. Insoluble in ether.
OCC(O)C(O)C(O)C(O)CO
(2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
6251
182.17
D-Mannitol
165°C to 167°C
290°C to 295°C (3.5 mmHg)
C6H14O6
1721898
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
FBPFZTCFMRRESA-UHFFFAOYNA-N
+24° (in Borax Solution)
182.17
CHEBI:16899
97+%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-711-8

RTECSNumber : OP2060000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only