missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Mannitol, 97+%

Catalog No. AAA140300B
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA140300B 1000 g
AAA1403030 250 g
AAA140300I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA140300B Supplier Thermo Scientific Chemicals Supplier No. A140300B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69-65-8 | C6H14O6 | 182.17 g/mol

D-Mannitol is used in the determination of glomerular filtration rate. It is used as an inert osmotic control substance. Further, it is used in the treatment of oliguric renal failure. It is also used as a sweetener for people with diabetes, in chewing gums and acts as an adulterant for heroin, methamphetamines and cocaine. It finds application as a research tool in cell biological studies, usually to control osmolarity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Mannitol is used in the determination of glomerular filtration rate. It is used as an inert osmotic control substance. Further, it is used in the treatment of oliguric renal failure. It is also used as a sweetener for people with diabetes, in chewing gums and acts as an adulterant for heroin, methamphetamines and cocaine. It finds application as a research tool in cell biological studies, usually to control osmolarity.

Solubility
Soluble in water, alcohol, aniline, pyridine and glycerol. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
IUPAC Name (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Melting Point 165°C to 167°C
Density 1.489
Boiling Point 290°C to 295°C (3.5 mmHg)
Odor Odorless
Quantity 1000 g
Beilstein 1721898
Merck Index 14,5745
Solubility Information Soluble in water,alcohol,aniline,pyridine and glycerol. Insoluble in ether.
Optical Rotation +24° (in Borax Solution)
Formula Weight 182.17
Percent Purity 97+%
Chemical Name or Material D-Mannitol
Show More Show Less
EINECSNumber 200-711-8
RTECSNumber OP2060000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only