missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Mannitol, 97+%

CAS: 69-65-8 | C6H14O6 | 182.17 g/mol

Supplier:  Thermo Scientific Chemicals A140300I

Quantity 5000 g

 View more versions of this product

Catalog No. AAA140300I


Only null left

Description

Description

D-Mannitol is used in the determination of glomerular filtration rate. It is used as an inert osmotic control substance. Further, it is used in the treatment of oliguric renal failure. It is also used as a sweetener for people with diabetes, in chewing gums and acts as an adulterant for heroin, methamphetamines and cocaine. It finds application as a research tool in cell biological studies, usually to control osmolarity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Mannitol is used in the determination of glomerular filtration rate. It is used as an inert osmotic control substance. Further, it is used in the treatment of oliguric renal failure. It is also used as a sweetener for people with diabetes, in chewing gums and acts as an adulterant for heroin, methamphetamines and cocaine. It finds application as a research tool in cell biological studies, usually to control osmolarity.

Solubility
Soluble in water, alcohol, aniline, pyridine and glycerol. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

69-65-8
182.17
FBPFZTCFMRRESA-UHFFFAOYNA-N
6251
(2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
C6H14O6
MFCD00064287
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
CHEBI:16899
OCC(O)C(O)C(O)C(O)CO

Specifications

D-Mannitol
165°C to 167°C
290°C to 295°C (3.5 mmHg)
C6H14O6
MFCD00064287
14,5745
Soluble in water,alcohol,aniline,pyridine and glycerol. Insoluble in ether.
OCC(O)C(O)C(O)C(O)CO
(2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
6251
182.17
69-65-8
1.489
Odorless
5000 g
1721898
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
FBPFZTCFMRRESA-UHFFFAOYNA-N
+24° (in Borax Solution)
182.17
CHEBI:16899
97+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-711-8

RTECSNumber : OP2060000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions