missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Lyxose 98.0+%, TCI America™

Catalog No. L00735G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L00735G 5 g
L00731G 1 g
L007325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L00735G Supplier TCI America Supplier No. L00735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1114-34-7
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00064362
InChI Key PYMYPHUHKUWMLA-UOWFLXDJSA-N
Synonym 2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci
PubChem CID 65550
ChEBI CHEBI:16789
IUPAC Name (2S,3S,4R)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 150.13
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-Lyxose