missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ D-Lysine hydrochloride, 99+%
Supplier: Thermo Scientific™ 243080100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
| D-Lysine hydrochloride | |
| 1ppm max. | |
| Authentic | |
| 99+% | |
| C6H15ClN2O2 | |
| H2N(CH2)4CH(NH2)COOH·HCl | |
| -20.5° to -21.5° (20°C, 589nm) (c=8, 6N HCl) | |
| d-lysine hydrochloride, d-lysine monohydrochloride, d-lysine hcl, d-lysine, monohydrochloride, h-d-lys-oh.hcl, d-lysine, hydrochloride, unii-kk611jde1n, lysine, monohydrochloride, d, r-2,6-diaminohexanoic acid hydrochloride, lysine, hydrochloride, d | |
| BVHLGVCQOALMSV-NUBCRITNSA-N | |
| (2R)-2,6-diaminohexanoic acid;hydrochloride | |
| 182.648 | |
| 81691 | |
| 182.65 |
| 7274-88-6 | |
| 10ppm max. | |
| 30ppm max. | |
| 0.3% max. (105°C, 3 hrs) | |
| Glass bottle | |
| MFCD00012920 | |
| 01,959; 07,79; 01,977 | |
| 0.1% max. | |
| C(CCN)CC(C(=O)O)N.Cl | |
| −21° (20°C c=8,6N HCl) | |
| 10g | |
| CHEBI:53558 | |
| 99.0 to 101.0% |