missing translation for 'onlineSavingsMsg'
Learn More

D-Leucine 98.0+%, TCI America™

Catalog No. L00275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L00275G 5 g
L002725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L00275G Supplier TCI America Supplier No. L00275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 328-38-1
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00063088
InChI Key ROHFNLRQFUQHCH-RXMQYKEDSA-N
Synonym d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin
PubChem CID 439524
ChEBI CHEBI:28225
IUPAC Name (2R)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@@H](N)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 131.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Leucine
EINECSNumber (9)-1632
RTECSNumber OH2840000
TSCA Yes