missing translation for 'onlineSavingsMsg'
Learn More

D-Leucine, 99%, Thermo Scientific Chemicals

Catalog No. AAA1484214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1484214 25 g
AAA1484206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1484214 Supplier Thermo Scientific Chemicals Supplier No. A1484214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

D-Leucine is used in the formation of sterols. It is also used as a selectable marker.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Leucine is used in the formation of sterols. It is also used as a selectable marker.

Solubility
Soluble in water and hydrochloric acid.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 328-38-1
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00063088
InChI Key ROHFNLRQFUQHCH-RXMQYKEDSA-N
Synonym d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzin
PubChem CID 439524
ChEBI CHEBI:28225
IUPAC Name (2R)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@@H](N)C(O)=O

Specifications

Melting Point >300°C
Beilstein 1721721
Solubility Information Soluble in water and hydrochloric acid.
Quantity 25 g
Formula Weight 131.18
Optical Rotation −15° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material D-Leucine
EINECSNumber 206-327-7
RTECSNumber OH2840000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only