missing translation for 'onlineSavingsMsg'
Learn More

D-Isosorbide, 98%

Catalog No. AAA13989D9
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
5 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA13989D9 5 x 1 kg
AAA1398918 50 g
AAA1398930 250 g
AAA139890B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA13989D9 Supplier Thermo Scientific Chemicals Supplier No. A13989D9
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 652-67-5 | C6H10O4 | 146.142 g/mol

D-Isosorbide is used as a reagent in the preparation of detergents, agrochemicals, cleansers, cosmetics and vasodilators. It is also employed as a pharmaceutical intermediate for dimethyl ether and mono- and dinitrate derivatives, which is used as vasodilators and coronary disease therapeutics. It is used in the treatment of hydrocephalus and glaucoma. It finds application as a building block for polymers like polycarbonates and polyesters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Isosorbide is used as a reagent in the preparation of detergents, agrochemicals, cleansers, cosmetics and vasodilators. It is also employed as a pharmaceutical intermediate for dimethyl ether and mono- and dinitrate derivatives, which is used as vasodilators and coronary disease therapeutics. It is used in the treatment of hydrocephalus and glaucoma. It finds application as a building block for polymers like polycarbonates and polyesters.

Solubility
Soluble in alcohols, water and ketones.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 652-67-5
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00064827
InChI Key KLDXJTOLSGUMSJ-JGWLITMVSA-N
Synonym isosorbide, isobide, devicoran, hydronol, ismotic, sorbid, +-d-isosorbide, vascardin dinitrate, 3r,3ar,6s,6ar-hexahydrofuro 3,2-b furan-3,6-diol, 1,4-dianhydrosorbitol
PubChem CID 12597
IUPAC Name (3S,3aR,6R,6aR)-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-3,6-diol
SMILES C1C(C2C(O1)C(CO2)O)O

Specifications

Melting Point 56°C to 64°C
Quantity 5 x 1 kg
Odor Odorless
Beilstein 80510
Merck Index 14,5224
Solubility Information Soluble in alcohols,water and ketones.
Formula Weight 146.14
Percent Purity 98%
Chemical Name or Material D-Isosorbide
EINECSNumber 211-492-3
RTECSNumber LZ4380000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only