missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Isoleucine, 98%

Catalog No. AAH27488MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27488MD 250 mg
AAH2748803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27488MD Supplier Thermo Scientific Chemicals Supplier No. H27488MD
Only null left

CAS: 319-78-8 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 319-78-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064221
InChI Key AGPKZVBTJJNPAG-LXDRQGDMNA-N
Synonym d-isoleucine, 2r,3r-2-amino-3-methylpentanoic acid, h-d-ile-oh, isoleucine, d, unii-v87gja0g54, r-isoleucine, 2r,3r-2-amino-3-methylvaleric acid, alloisoleucin, d-ile, ambotzhaa1186
PubChem CID 76551
ChEBI CHEBI:27730
IUPAC Name (2R,3R)-2-amino-3-methylpentanoic acid
SMILES CC[C@@H](C)[C@@H](N)C(O)=O

Specifications

Melting Point 265°C (sublimation)
Quantity 250 mg
Formula Weight 131.18
Optical Rotation −40° (c=5 in 5N HCl)
Percent Purity 98%
Chemical Name or Material D-Isoleucine
EINECSNumber 206-269-2
RTECSNumber NR4700000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only