missing translation for 'onlineSavingsMsg'
Learn More

D-Homophenylalanine, 98%

Catalog No. AAH2762906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2762906 5 g
AAH2762903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2762906 Supplier Thermo Scientific Chemicals Supplier No. H2762906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 82795-51-5 | C10H13NO2 | 179.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82795-51-5
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00063091
InChI Key JTTHKOPSMAVJFE-SECBINFHSA-N
Synonym d-homophenylalanine, h-d-hophe-oh, 2r-2-amino-4-phenylbutanoic acid, r-2-amino-4-phenylbutanoic acid, d-homophenyl-ala, h-d-homophe-oh, r-homophenylalanine, r-2-amino-4-phenylbutyric acid, unii-5847zw3pep, d-homophenyl alanine
PubChem CID 6950302
IUPAC Name (2R)-2-amino-4-phenylbutanoic acid
SMILES N[C@H](CCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point >300°C
Beilstein 4675530
Quantity 5 g
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material D-Homophenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only