missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Glutamine 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 146.15 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | D-Glutamine |
Safety and Handling
| TSCA | No |