missing translation for 'onlineSavingsMsg'
Learn More

D-Glutamic Acid 98.0+%, TCI America™

Catalog No. G005725G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G005725G 25 g
G0057250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G005725G Supplier TCI America Supplier No. G005725G

Chemical Identifiers

CAS 6893-26-1
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00063112
InChI Key WHUUTDBJXJRKMK-GSVOUGTGSA-N
Synonym d-glutamic acid, h-d-glu-oh, d-glu, r-2-aminopentanedioic acid, 2r-2-aminopentanedioic acid, d-glutamate, d-2-aminopentanedioic acid, glutamic acid d-form, d--glutamic acid, d-glutaminic acid
PubChem CID 23327
ChEBI CHEBI:15966
IUPAC Name (2R)-2-aminopentanedioic acid
SMILES C(CC(=O)O)C(C(=O)O)N

Specifications

Color White
Quantity 25 g
Formula Weight 147.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-Glutamic Acid
EINECSNumber (9)-1573
TSCA Yes