missing translation for 'onlineSavingsMsg'
Learn More

D-Glucuronic acid, 98+%

Catalog No. AAL1435006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1435006 5 g
AAL1435014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1435006 Supplier Thermo Scientific Chemicals Supplier No. L1435006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6556-12-3 | C6H10O7 | 194.14 g/mol

D-Glucuronic acid is used as pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glucuronic acid is used as pharmaceutical intermediate and in chemical research.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 6556-12-3
Molecular Formula C6H10O7
Molecular Weight (g/mol) 194.14
MDL Number MFCD00077778
InChI Key IAJILQKETJEXLJ-QTBDOELSSA-N
Synonym 2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoic acid, aldehydo-d-glucuronic acid, unii-8a5d83q4rw, rel, d-glucuronic acid, d-+ glucuronic acid, d03jeu, d-glucuronic acid 5g
PubChem CID 65041
ChEBI CHEBI:47953
IUPAC Name (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
SMILES C(=O)C(C(C(C(C(=O)O)O)O)O)O

Specifications

Melting Point ∽158°C (decomposition)
Quantity 5 g
Beilstein 1727083
Merck Index 14,4465
Solubility Information Soluble in water.
Optical Rotation +35° (c=6 in Water)
Formula Weight 194.14
Percent Purity ≥98%
Chemical Name or Material D-Glucuronic acid
EINECSNumber 229-486-4
RTECSNumber LZ8836600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only