missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Glucose, anhydrous, 99%

CAS: 50-99-7 | C6H12O6 | 180.156 g/mol

Supplier:  Thermo Scientific Chemicals A168280C

Quantity 10,000 g

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA168280C


Il en reste null

Description

Description

D-Glucose is used in various metabolic processes including enzymic synthesis of cyclohexyl-alpha and beta -D-glucosides. It is involved in the detection of type 2 diabetes mellitus and the most widely used aldohexose in living organisms. Its availability influences psychological processes since it is a primary energy source for the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glucose is used in various metabolic processes including enzymic synthesis of cyclohexyl-alpha and beta -D-glucosides. It is involved in the detection of type 2 diabetes mellitus and the most widely used aldohexose in living organisms. Its availability influences psychological processes since it is a primary energy source for the brain.

Solubility
Soluble in water, hot glacial acetic acid, pyridine, aniline and dimethyl sulfoxide. Slightly soluble in methanol, ethanol and acetone.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
Spécifications

Identifiants chimiques

50-99-7
180.156
GZCGUPFRVQAUEE-SLPGGIOYSA-N
107526
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
C6H12O6
MFCD00063774
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
CHEBI:42758
C(C(C(C(C(C=O)O)O)O)O)O

Spécifications

D-(+)-Glucose
50-99-7
99%
10,000 g
1281608
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
GZCGUPFRVQAUEE-SLPGGIOYSA-N
C(C(C(C(C(C=O)O)O)O)O)O
180.156
CHEBI:42758
99%
anhydrous
147°C to 153°C
C6H12O6
MFCD00063774
14,4459
Soluble in water,hot glacial acetic acid,pyridine,aniline and dimethyl sulfoxide. Slightly soluble in methanol,ethanol and acetone.
+53° (c=10 in Water, 3h)
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
107526
180.16
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

EINECSNumber : 200-075-1

RTECSNumber : LZ6600000

TSCA : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions