missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Glucosamine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 194°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 215.63 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | D-(+)-Glucosamine Hydrochloride |
Safety and Handling
| RTECSNumber | LZ6665000 |
| TSCA | Yes |