missing translation for 'onlineSavingsMsg'
Learn More

D-Gluconic acid, 50% aq. soln.

Catalog No. AAA16806AP
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16806AP 500 mL
AAA168060F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA16806AP Supplier Thermo Scientific Chemicals Supplier No. A16806AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 526-95-4 | C6H12O7 | 196.155 g/mol

Gluconic acid occurs naturally in fruit, honey, and wine. As a food additive it is an acidity regulator. It is also used in cleaning products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gluconic acid occurs naturally in fruit, honey, and wine. As a food additive it is an acidity regulator. It is also used in cleaning products.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 526-95-4
Molecular Formula C6H12O7
Molecular Weight (g/mol) 196.155
MDL Number MFCD00004240
InChI Key RGHNJXZEOKUKBD-SQOUGZDYSA-N
Synonym gluconic acid, d-gluconic acid, dextronic acid, maltonic acid, glycogenic acid, glosanto, gluconate, pentahydroxycaproic acid, gluconic acid, d, d-gluconate
PubChem CID 10690
ChEBI CHEBI:33198
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
SMILES C(C(C(C(C(C(=O)O)O)O)O)O)O

Specifications

Density 1.234
Quantity 500 mL
Optical Rotation +10° (Neat)
Refractive Index 1.418
UN Number UN3265
Beilstein 1726055
Merck Index 144456
Solubility Information Soluble in water.
Formula Weight 196.16 (Anhydrous)
Concentration or Composition (by Analyte or Components) 50% aq. soln.
Chemical Name or Material D-Gluconic acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 208-401-4
RTECSNumber LZ5057100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only