missing translation for 'onlineSavingsMsg'
Learn More

D-Glucal 97.0+%, TCI America™

Catalog No. p-7130219
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02741G 1 g
G02745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02741G Supplier TCI America Supplier No. G02741G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13265-84-4
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00067186
InChI Key YVECGMZCTULTIS-PBXRRBTRSA-N
Synonym d-glucal, 2r,3s,4r-2-hydroxymethyl-3,4-dihydro-2h-pyran-3,4-diol, glucal, d-arabino-hex-1-enitol, 1,5-anhydro-2-deoxy, 1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, dgo, pubchem10547, ksc921e7l, 1,2-dideoxy-d-arabino-1-hexenopyranose
PubChem CID 2734736
IUPAC Name (2R,3S,4R)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol
SMILES C1=COC(C(C1O)O)CO

Specifications

Melting Point 62°C
Color White
Quantity 1 g
Formula Weight 146.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material D-Glucal
TSCA No
Recommended Storage Refrigerator