missing translation for 'onlineSavingsMsg'
Learn More

D-gamma-Tocopherol, 95%

Catalog No. AC421041000
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421041000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC421041000 Supplier Thermo Scientific Chemicals Supplier No. 421041000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54-28-4 | C28H48O2 | 416.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54-28-4
Molecular Formula C28H48O2
Molecular Weight (g/mol) 416.69
MDL Number MFCD00066529
InChI Key QUEDXNHFTDJVIY-DQCZWYHMSA-N
Synonym gamma-tocopherol, +-gamma-tocopherol, d-gamma-tocopherol, 7,8-dimethyltocol, unii-8ef1z1238f, rrr-gamma-tocopherol, r,r,r-gamma-tocopherol, 2r-2,7,8-trimethyl-2-4r,8r-4,8,12-trimethyltridecyl chroman-6-ol, 2r-2,7,8-trimethyl-2-4r,8r-4,8,12-trimethyltridecyl-3,4-dihydro-2h-chromen-6-ol, 2r-3,4-dihydro-2,7,8-trimethyl-2-4r,8r-4,8,12-trimethyltridecyl-2h-1-benzopyran-6-ol
PubChem CID 92729
ChEBI CHEBI:18185
IUPAC Name (2R)-2,7,8-trimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
SMILES CC1=C(C=C2CCC(OC2=C1C)(C)CCCC(C)CCCC(C)CCCC(C)C)O

Specifications

Color Brown
Boiling Point 200°C (0.1 mmHg)
Flash Point >93°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 15, 9655
Specific Rotation Condition +30° (25°C c=2.00 in isooctane)
Specific Rotation + 30
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,acetone,chloroform,ether,,oils,fats
Formula Weight 416.69
Percent Purity 95%
Physical Form Viscous Liquid
Chemical Name or Material D-γ-Tocopherol, 95%
Show More Show Less
EINECSNumber 200-201-5

RUO – Research Use Only