missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Fucose 98.0+%, TCI America™

Catalog No. D00495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00495G 5 g
D00491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D00495G Supplier TCI America Supplier No. D00495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3615-37-0
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.157
MDL Number MFCD00135603
InChI Key PNNNRSAQSRJVSB-DPYQTVNSSA-N
Synonym 2r,3s,4s,5r-2,3,4,5-tetrahydroxyhexanal, aldehydo-d-fucose, d +-fucose, galactose, 6-deoxy, d-galactose, 6-deoxy, fucose, d, +-fucose, d0o8ie, d-+-fucose, d-+-fucose;6-deoxy-d-galactose
PubChem CID 94270
ChEBI CHEBI:48203
IUPAC Name (2R,3S,4S,5R)-2,3,4,5-tetrahydroxyhexanal
SMILES CC(C(C(C(C=O)O)O)O)O

Specifications

Melting Point 137°C
Color White
Quantity 5 g
Formula Weight 164.16
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Fucose