missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Fructose 99.0+%, TCI America™

Catalog No. F0060500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0060500G 500 g
F006025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0060500G Supplier TCI America Supplier No. F0060500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00148910
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
Synonym d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Color White
Quantity 500 g
Formula Weight 180.16
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-Fructose
EINECSNumber (8)-0047
RTECSNumber LS7120000
TSCA Yes