missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Dextrose, (Crystalline), MP Biomedicals
Chemical Identifiers
| CAS | 50-99-7 |
|---|---|
| Molecular Formula | C6H12O6 |
| Molecular Weight (g/mol) | 180.156 |
| InChI Key | GZCGUPFRVQAUEE-SLPGGIOYSA-N |
| Synonym | 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form |
| PubChem CID | 107526 |
| ChEBI | CHEBI:42758 |
| IUPAC Name | (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
| SMILES | C(C(C(C(C(C=O)O)O)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN90152101
|
N/A | 1 kg |
N/A
|
|
|||||
|
ICN90152105
|
N/A | 5 kg |
N/A
|
|
|||||
|
ICN90152110
|
N/A | 10 kg |
N/A
|
|
|||||
|
ICN90152125
|
N/A | 25 kg |
N/A
|
|
|||||
Chemical Identifiers
| 50-99-7 | |
| 180.156 | |
| 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
| CHEBI:42758 | |
| C(C(C(C(C(C=O)O)O)O)O)O |
| C6H12O6 | |
| GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
| 107526 | |
| (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
Specifications
| 50-99-7 | |
| 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
| C(C(C(C(C(C=O)O)O)O)O)O | |
| 180.156 | |
| CHEBI:42758 | |
| Crystalline |
| C6H12O6 | |
| GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
| (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal | |
| 107526 | |
| 180.2 | |
| D-(+)-Dextrose |
Safety and Handling
Recommended Storage : Room Temperature