missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Dextrose, (Crystalline), MP Biomedicals

Catalog No. ICN90152125
Change view
Click to view available options
Quantity:
1 kg
5 kg
10 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN90152125 25 kg
ICN90152101 1 kg
ICN90152105 5 kg
ICN90152110 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN90152125 Supplier MP Biomedicals Inc Supplier No. 0290152125

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Quantity 25 kg
Formula Weight 180.2
Physical Form Crystalline
Chemical Name or Material D-(+)-Dextrose
Recommended Storage Room Temperature