missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Dextrose, (Crystalline), MP Biomedicals
Specifications
Specifications
| Quantity | 10 kg |
| Formula Weight | 180.2 |
| Physical Form | Crystalline |
| Chemical Name or Material | D-(+)-Dextrose |
Safety and Handling
| Recommended Storage | Room Temperature |