missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Cycloserine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 137°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 102.09 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | D-(+)-Cycloserine |
Safety and Handling
| RTECSNumber | NY2975000 |
| TSCA | No |
| Recommended Storage | Refrigerator |