Learn More
Abcam Inc D-(+)-Cellobiose, beta-glucosidase substrate, 25G
Numéro de catalogue. 502704663
Encompass
MW 342.3 g/mol, Purity >99%. Disaccharide consisting of two -glucose molecules linked by a -1,4 glycosidic bond. Used as a substrate of -glucosidase.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502704663 Fournisseur Abcam Inc Code fournisseur AB14639125G
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 342.3 g/mol, Purity >99%. Disaccharide consisting of two -glucose molecules linked by a -1,4 glycosidic bond. Used as a substrate of -glucosidase.
The product is subject to the following: Abcam Restricted Use Statement