missing translation for 'onlineSavingsMsg'
Learn More

D-Aspartic acid dimethyl ester hydrochloride, 98%

Catalog No. AAH6319706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6319706 5 g
AAH6319703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6319706 Supplier Thermo Scientific Chemicals Supplier No. H6319706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69630-50-8 | C6H12ClNO4 | 197.615 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69630-50-8
Molecular Formula C6H12ClNO4
Molecular Weight (g/mol) 197.615
MDL Number MFCD00070384
InChI Key PNLXWGDXZOYUKB-PGMHMLKASA-N
Synonym d-aspartic acid dimethyl ester hydrochloride, h-d-asp ome-ome.hcl, h-d-asp ome-oh hcl, r-dimethyl 2-aminosuccinate hydrochloride, d-aspartic acid, dimethyl ester, hydrochloride, d-aspartic acid-1,4-dimethyl ester hydrochloride, 1,4-dimethyl 2r-2-aminobutanedioate hydrochloride, h-d-asp diome . hcl, h-d-asp ome-ome hcl, d-aspartic acid dimethyl ester hcl
PubChem CID 51051594
IUPAC Name dimethyl (2R)-2-aminobutanedioate;hydrochloride
SMILES COC(=O)CC(C(=O)OC)N.Cl

Specifications

Melting Point 114°C to 119°C
Quantity 5 g
Formula Weight 197.62
Percent Purity 98%
Physical Form Solid
Chemical Name or Material D-Aspartic acid dimethyl ester hydrochloride
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only