missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Arginine monohydrochloride, 98%

Catalog No. AAA1622214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1622214 25 g
AAA1622203 1 g
AAA1622206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1622214 Supplier Thermo Scientific Chemicals Supplier No. A1622214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 627-75-8 | C6H15ClN4O2 | 210.662 g/mol

The influence in creatinine metabolismis that by that d-arginine monohydrochloride caused slightly larger increases in muscle creatine. D-Arginyl ethyl ester was synthesized by vigorously stirring D-arginine monohydrochloride and excess anhydrous ethanol in an ice bath.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The influence in creatinine metabolismis that by that d-arginine monohydrochloride caused slightly larger increases in muscle creatine. D-Arginyl ethyl ester was synthesized by vigorously stirring D-arginine monohydrochloride and excess anhydrous ethanol in an ice bath.

Solubility
Soluble in water (50 mg/ml), and 3N HCl (5).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 627-75-8
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
MDL Number MFCD00012620
InChI Key KWTQSFXGGICVPE-PGMHMLKASA-N
Synonym d-arginine hydrochloride, h-d-arg-oh.hcl, d-arginine monohydrochloride, d-arginine, monohydrochloride, unii-aw80ygd17d, d-arginine hcl, arginine, monohydrochloride, d, aw80ygd17d, arginine, chloride, d--arginine hydrochloride
PubChem CID 12326
IUPAC Name (2R)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Melting Point 216°C to 218°C
Beilstein 5773432
Solubility Information Soluble in water (50mg/ml),and 3N HCl (5).
Quantity 25 g
Formula Weight 210.67
Percent Purity 99%
Sensitivity Hygroscopic
Chemical Name or Material D-Arginine monohydrochloride
EINECSNumber 211-010-1
RTECSNumber CF1995000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only