missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Arginine, 98%

Catalog No. AAA1613706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1613706 5 g
AAA1613703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1613706 Supplier Thermo Scientific Chemicals Supplier No. A1613706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 157-06-2 | C6H14N4O2 | 174.204 g/mol

1-Deamino-8-D-arginine vasopressin makes a new pharmacological approach to the management of haemophilia and von Willebrand's disease. A novel nonenzymatic pathway for the generation of nitric oxide is by the reaction of hydrogen peroxide and d- or l-arginine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Deamino-8-D-arginine vasopressin makes a new pharmacological approach to the management of haemophilia and von Willebrand’s disease. A novel nonenzymatic pathway for the generation of nitric oxide is by the reaction of hydrogen peroxide and d- or l-arginine.

Solubility
Insoluble in water.

Notes
Air Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Chemical Identifiers

CAS 157-06-2
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.204
MDL Number MFCD00063116
InChI Key ODKSFYDXXFIFQN-SCSAIBSYSA-N
Synonym d-arginine, h-d-arg-oh, r-2-amino-5-guanidinopentanoic acid, d-arginin, d-arg, d---arginine, arginine, d, d-2-amino-5-guanidinovaleric acid, d--arginine, unii-r54z304z7c
PubChem CID 71070
ChEBI CHEBI:15816
IUPAC Name (2R)-2-amino-5-(diaminomethylideneamino)pentanoic acid
SMILES C(CC(C(=O)O)N)CN=C(N)N

Specifications

Melting Point ∼235°C
Beilstein 1725412
Solubility Information Soluble in water at 148g/L.
Quantity 5 g
Formula Weight 174.2
Sensitivity Air Sensitive
Percent Purity 98%
Chemical Name or Material D-Arginine
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 205-866-5
RTECSNumber CF1934220
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only