missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Arabinose 99.0+%, TCI America™

Supplier:  TCI America A051325G

 View more versions of this product

Catalog No. A051325G


Only null left

Description

Description

Specifications

Specifications

D-(-)-Arabinose
161°C
MFCD00135608
d---arabinose, d-arabinose, dl-arabinose, arabinose, d, arabinose,d, 2s,3r,4r-2,3,4,5-tetrahydroxypentanal, unii-f0w6etz4e5, --arabinose, aldehydo-d-arabinose, f0w6etz4e5
C(C(C(C(C=O)O)O)O)O
150.13
CHEBI:46983
≥99.0% (HPLC)
10323-20-3
C5H10O5
25 g
PYMYPHUHKUWMLA-WDCZJNDASA-N
(2S,3R,4R)-2,3,4,5-tetrahydroxypentanal
66308
150.13
Crystalline Powder

Chemical Identifiers

10323-20-3
150.13
PYMYPHUHKUWMLA-WDCZJNDASA-N
66308
(2S,3R,4R)-2,3,4,5-tetrahydroxypentanal
C5H10O5
MFCD00135608
d---arabinose, d-arabinose, dl-arabinose, arabinose, d, arabinose,d, 2s,3r,4r-2,3,4,5-tetrahydroxypentanal, unii-f0w6etz4e5, --arabinose, aldehydo-d-arabinose, f0w6etz4e5
CHEBI:46983
C(C(C(C(C=O)O)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2470

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions