missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Arabinose 99.0+%, TCI America™

Catalog No. p-7116940
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A051325G 25 g
A0513100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A051325G Supplier TCI America Supplier No. A051325G
Only null left

Chemical Identifiers

CAS 10323-20-3
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00135608
InChI Key PYMYPHUHKUWMLA-WDCZJNDASA-N
Synonym d---arabinose, d-arabinose, dl-arabinose, arabinose, d, arabinose,d, 2s,3r,4r-2,3,4,5-tetrahydroxypentanal, unii-f0w6etz4e5, --arabinose, aldehydo-d-arabinose, f0w6etz4e5
PubChem CID 66308
ChEBI CHEBI:46983
IUPAC Name (2S,3R,4R)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O

Specifications

Melting Point 161°C
Quantity 25 g
Formula Weight 150.13
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-Arabinose
EINECSNumber (2)-2470
TSCA Yes