missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Alanine, 99+%

Catalog No. AC172080050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172080050 5 g Glass Bottle
AC172080250 25 g Glass Bottle
AC172081000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC172080050 Supplier Thermo Scientific Chemicals Supplier No. 172080050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 338-69-2 | C3H7NO2 | 89.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 338-69-2
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00008077
InChI Key QNAYBMKLOCPYGJ-UWTATZPHSA-N
Synonym d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form
PubChem CID 71080
ChEBI CHEBI:15570
IUPAC Name (2R)-2-aminopropanoic acid
SMILES C[C@@H](N)C(O)=O

Specifications

Melting Point 278.0°C to 282.0°C
Arsenic (As) 2ppm max. (As2O3)
Color White
Chloride 0.1% max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Loss on Drying 0.2% max. (105°C, 3 hrs)
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula CH3CH(NH2)CO2H
Specific Rotation − 14.50
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Sulfated Ash 0.2% max.
Solubility Information Solubility in water: 155g/L in water (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents
Quantity 5 g
Formula Weight 89.09
Percent Purity ≥99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D-Alanine
Show More Show Less

RUO – Research Use Only