Learn More
Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips)
Achieve ultimate performance in automated pipetting with these tips--superior quality with the flexibility to choose tips for any application.
$1368.65 - $10819.03
| Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Filtered | Packaging | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Filtered | Packaging | Price | Quantity | ||
|
14-387-674
|
Thermo Scientific™
MX5317 |
30 μL | Sterile | Non-filtered | 20 x 384 tips |
Case of 10 Each for $7,610.12
|
|
||
|
14-387-693
|
Thermo Scientific™
MX5318 |
30 μL | Sterile | Filtered | 20 x 384 tips |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
14-387-690
|
Thermo Scientific™
MX5328 |
100 μL | Sterile | Filtered | 20 x 384 tips |
N/A
|
N/A | ||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
14-387-699
|
Thermo Scientific™
5526 |
100 μL | Non-sterile | Non-filtered | 20 x 96 tips |
Case of 10 Each for $1,368.65
|
|
||
|
14-387-663
|
Thermo Scientific™
MX5528 |
100 μL | Sterile | Filtered | 20 x 96 tips |
Case of 10 Each for $2,476.94
|
|
||
Description
Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.
Features:- Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
- Highly polished molds and cores ensure low liquid retention.
- Small internal tip orifice provides better small volume droplet formation
- Sterile, sterile filtered, wide-bore, and extended-length tip options are available
- Tips are thoroughly inspected for proper formation and straightness
- Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
- Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.
Includes: 20 magazines per case
Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3 and PlateMate™ Plus Systems.