missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips)

Catalog No. 14387674
Change view
Click to view available options
Volume (Metric):
30 μL
100 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Packaging:
20 x 384 tips
20 x 96 tips
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Filtered Packaging
14387674 30 μL Sterile Non-filtered 20 x 384 tips
14387693 30 μL Sterile Filtered 20 x 384 tips
14387690 100 μL Sterile Filtered 20 x 384 tips
14387699 100 μL Non-sterile Non-filtered 20 x 96 tips
14387663 100 μL Sterile Filtered 20 x 96 tips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14387674 Supplier Thermo Scientific™ Supplier No. MX5317

Achieve ultimate performance in automated pipetting with these tips--superior quality with the flexibility to choose tips for any application.

Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.

Features:

  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access
  • Highly polished molds and cores ensure low liquid retention
  • Small internal tip orifice provides better small volume droplet formation
  • Sterile, sterile-filtered, wide-bore, and extended-length tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination

Includes: 20 magazines per case

Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3 and PlateMate™ Plus Systems.

Specifications

Volume (Metric) 30 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Length (English) Standard
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Rack
Tip Style Automation
Packaging 20 x 384 tips
Quantity 7680 tips
Disposable Single-use
For Use With (Equipment) Thermo Scientific Matrix Hydra DT, Thermo Scientific PlateMate Plus, Thermo Scientific Versette
Product Line D.A.R.T.S
Show More Show Less