missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-3-Phenyllactic acid, 98%

Catalog No. AAH3509606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3509606 5 g
AAH3509603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3509606 Supplier Thermo Scientific Chemicals Supplier No. H3509606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7326-19-4 | C9H10O3 | 166.18 g/mol

D-(+)-3-Phenyllactic acid is a chiral building block that is used in preparation of statine. Starting material in preparation of the hypoglycemic agent enlitazone and of (15)N-labeled phenylalanine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-3-Phenyllactic acid is a chiral building block that is used in preparation of statine. Starting material in preparation of the hypoglycemic agent enlitazone and of (15)N-labeled phenylalanine.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 7326-19-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00078062
InChI Key VOXXWSYKYCBWHO-MRVPVSSYSA-N
Synonym r-2-hydroxy-3-phenylpropionic acid, r-2-hydroxy-3-phenylpropanoic acid, 2r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-3-phenyllactic acid, r-phenyllactate, r-+-3-phenyllactic acid, 3-phenyl-d-lactic acid, r-3-phenyllactic acid, r-, a-hydroxy-3-phenylpropionic acid
PubChem CID 643327
ChEBI CHEBI:32978
IUPAC Name (2R)-2-hydroxy-3-phenylpropanoic acid
SMILES O[C@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 122°C to 127°C
Quantity 5 g
Solubility Information Soluble in water.
Optical Rotation +22° (c=1 in Water)
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material D-(+)-3-Phenyllactic acid
EINECSNumber 230-803-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only