missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-2-Phenylglycine ethyl ester hydrochloride, 98+%

Numéro de catalogue. AAL0156418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAL0156418 50 g
AAL0156409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAL0156418 Fournisseur Thermo Scientific Chemicals Code fournisseur L0156418
Il en reste null

CAS: 17609-48-2 | C10H14ClNO2 | 215.677 g/mol

D-(-)-2-Phenylglycine ethyl ester hydrochloride is used in pharmaceutical intermediates, in the combination of dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-2-Phenylglycine ethyl ester hydrochloride is used in pharmaceutical intermediates, in the combination of dyestuff.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Identifiants chimiques

CAS 17609-48-2
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.677
MDL Number MFCD00052005
InChI Key FNNXQLSKQSVNLL-SBSPUUFOSA-N
Synonym r-ethyl 2-amino-2-phenylacetate hydrochloride, r-ethyl 2-amino-2-phenylacetate hcl, ethyl 2r-2-amino-2-phenylacetate hydrochloride, d-phenylglycine-oet hcl, h-d-phg-oet . hcl, d---2-phenylglycine ethyl ester hydrochloride, d---alpha-phenylglycine ethyl ester hydrochloride, pubchem10931, d-phg-oet hydrochloride
PubChem CID 20833765
IUPAC Name ethyl (2R)-2-amino-2-phenylacetate;hydrochloride
SMILES CCOC(=O)C(C1=CC=CC=C1)N.Cl

Spécifications

Melting Point ∽190°C (decomposition)
Beilstein 5775455
Solubility Information Soluble in water.
Quantity 50 g
Formula Weight 215.68
Optical Rotation −94° (c=1 in Water)
Percent Purity ≥98%
Chemical Name or Material D-(-)-2-Phenylglycine ethyl ester hydrochloride
EINECSNumber 241-581-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only