missing translation for 'onlineSavingsMsg'
Learn More

D(+)-10-Camphorsulfonic acid, 99%

Catalog No. AC108225000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108225000 500 g Plastic bottle
AC108221000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108225000 Supplier Thermo Scientific Chemicals Supplier No. 108225000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3144-16-9 | C10H15O4S | 231.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3144-16-9
Molecular Formula C10H15O4S
Molecular Weight (g/mol) 231.29
MDL Number MFCD00064157,MFCD00074827
InChI Key MIOPJNTWMNEORI-XVKPBYJWSA-M
Synonym 1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
PubChem CID 65617
ChEBI CHEBI:55403
IUPAC Name [(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
SMILES CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2

Specifications

Melting Point >190.0°C
Color White
Loss on Drying 2% max. (1 g, 105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Beilstein 11,IV,642
Fieser 01,108; 02,58; 04,68; 13,62; 14,71; 16,58
Merck Index 15, 1736
Specific Rotation + 22.00
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in glacial acetic acid,slightly soluble in ethyl acetate,practically insoluble in ether,very soluble in ethanol
Formula Weight 232.3
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-10-Camphorsulfonic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 221-554-1
RTECSNumber ED1550000
TSCA TSCA

RUO – Research Use Only