missing translation for 'onlineSavingsMsg'
Learn More

Cytosine, 99+%

Catalog No. AC161761000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161761000 100 g Glass bottle
AC161760050 5 g Glass bottle
AC161760250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC161761000 Supplier Thermo Scientific Chemicals Supplier No. 161761000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71-30-7 | C4H5N3O | 111.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71-30-7
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD00006034
InChI Key OPTASPLRGRRNAP-UHFFFAOYSA-N
Synonym cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine
PubChem CID 597
ChEBI CHEBI:16040
SMILES NC1=CC=NC(=O)N1

Specifications

Melting Point >300.0°C
Color White to Yellow
Loss on Drying 0.5% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 24,314
Merck Index 15,2792
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alkalines and acids,slightly soluble in ethanol,chloroform,,insoluble in ether and hydrocarbons
Spectrophotometry 98% min. (lambda max. 275nm,pH 1) molar absorptivity 10400+/-2%,on dry basis
IUPAC Name 6-amino-1,2-dihydropyrimidin-2-one
Formula Weight 111.1
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Cytosine
Show More Show Less
EINECSNumber 200-749-5

RUO – Research Use Only