missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytosine, 98+%

Catalog No. AAA1473122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1473122 100 g
AAA1473106 5 g
AAA1473114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1473122 Supplier Thermo Scientific Chemicals Supplier No. A1473122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71-30-7 | C4H5N3O | 111.10 g/mol

Cytosine is a nucleoside used for proteomics research. It is also used as an enzyme substrate or precursor of effector molecules such as cytosine sugars.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytosine is a nucleoside used for proteomics research. It is also used as an enzyme substrate or precursor of effector molecules such as cytosine sugars.

Solubility
Clear to very slightly hazy colorless to faint yellow solution at 50 mg/ml in 0.5 M HCL.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 71-30-7
Molecular Formula C4H5N3O
Molecular Weight (g/mol) 111.10
MDL Number MFCD00006034
InChI Key OPTASPLRGRRNAP-UHFFFAOYSA-N
Synonym cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine
PubChem CID 597
ChEBI CHEBI:16040
SMILES NC1=CC=NC(=O)N1

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 2637
Merck Index 14,2795
Solubility Information Clear to very slightly hazy colorless to faint yellow solution at 50mg/ml in 0.5 M HCL.
IUPAC Name 6-amino-1,2-dihydropyrimidin-2-one
Formula Weight 111.1
Percent Purity ≥98%
Chemical Name or Material Cytosine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 200-749-5
RTECSNumber UW7350150
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only