missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cytochalasin D

Catalog No. 12331
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12331 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12331 Supplier R&D Systems™ Supplier No. 1233/1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Disrupts actin filament function

Potent disruptor of actin filament function. Alters tight junction permeability. Unlike cytochalasin B (Cat. No. 5474), does not inhibit monosaccharide transport across the plasma membrane.

Chemical Identifiers

CAS 22144-77-0
Molecular Formula C30H37NO6

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 507.63
Purity >95%
Molecular Weight (g/mol) 507.627
Chemical Name or Material Cytochalasin D
Recommended Storage Desiccate at -20°C