CAS RN 22144-77-0
IUPAC Name:
(3S,3aR,4S,6S,6aR,10S,12R,15R,15aR)-3-benzyl-6,12-dihydroxy-4,10,12-trimethyl-5-methylidene-1,11-dioxo-1H,2H,3H,3aH,4H,5H,6H,6aH,9H,10H,11H,12H,15H-cycloundeca[e]isoindol-15-yl acetate
Synonyms:
cytochalasin D
Molecular Weight (g/mol):
507.63
Molecular Formula:
C30H37NO6
InChi Key:
SDZRWUKZFQQKKV-JHADDHBZSA-N
SMILES:
C[C@H]1[C@H]2[C@H](CC3=CC=CC=C3)NC(=O)[C@@]22[C@@H](\C=C\C[C@H](C)C(=O)[C@](C)(O)\C=C\[C@H]2OC(C)=O)[C@H](O)C1=C