missing translation for 'onlineSavingsMsg'
Learn More

Cytidine 5'-Diphosphocholine Sodium Salt 98.0+%, TCI America™

Catalog No. C34455G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34455G 5 g
C34451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34455G Supplier TCI America Supplier No. C34455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33818-15-4
Molecular Formula C14H25N4NaO11P2
Molecular Weight (g/mol) 510.31
MDL Number MFCD00037486
InChI Key YWAFNFGRBBBSPD-OCMLZEEQSA-M
Synonym citicolinesodium, cytidine 5'-diphosphocholine sodium, cdp-choline sodium, citicoline, sodium, cdp-choline, sodium, cytidine 5'-diphosphocholine sodium salt, cytidine 5'-diphosphocholine sodium salt dihydrate, cytidine-5'-diphosphocholine monosodium salt, sodium 2s,3r,4s,5s-5-4-amino-2-oxo-1-pyrimidinyl-3,4-dihydroxy-2-oxolanyl methoxy-oxidophosphoryl 2-trimethylammonio ethyl phosphate, sodium 2s,3r,4s,5s-5-4-azanyl-2-oxidanylidene-pyrimidin-1-yl-3,4-bis oxidanyl oxolan-2-yl methoxy-oxidanidyl-phosphoryl 2-trimethylazaniumyl ethyl phosphate
PubChem CID 46738085
IUPAC Name sodium 2-(trimethylazaniumyl)ethyl ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
SMILES [Na+].C[N+](C)(C)CCOP([O-])(=O)OP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(N)=NC1=O

Specifications

Melting Point 250°C
Color White
Quantity 5 g
Formula Weight 510.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cytidine 5′-Diphosphocholine Sodium Salt
EINECSNumber (9)-0508
TSCA No
Recommended Storage Freezer