Learn More
Abcam Inc Cytidine 5'-diphosphate (CDP) sodium salt hydrate, CDP kinase substrate, 500MG
Numéro de catalogue. 502704838
Encompass
MW 469.18 g/mol, Purity >98%. CDP (nucleoside diphosphate) kinase substrate to produce CTP in support of DNA and RNA biosynthesis. Ribonucleotide reductase substrate to produce dCMP.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502704838 Fournisseur Abcam Inc Code fournisseur AB146214500MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 469.18 g/mol, Purity >98%. CDP (nucleoside diphosphate) kinase substrate to produce CTP in support of DNA and RNA biosynthesis. Ribonucleotide reductase substrate to produce dCMP.
The product is subject to the following: Abcam Restricted Use Statement