missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Cytation 5 Cell Imaging Multi-Mode Reader
GREENER_CHOICE

Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. For Research Use Only. Not for use in diagnostic procedures.

Supplier:  Agilent BioTek CYT5MWSN

 Afficher plus de versions de ce produit

Numéro de catalogue. BTCYT5MW

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

Cytation 5 combines automated microscopy and conventional microplate detection in a configurable, upgradable platform. The microscopy module offers up to 60x magnification in fluorescence, brightfield, high contrast brightfield, color brightfield, and phase contrast to address many applications and workflows

The multi-mode detection modules include filter- and monochromator-based fluorescence detection, luminescence, and UV-Vis absorbance detection. Gen5 software provides complete control over all imaging and data capture, plus powerful image and data analysis.

Spécifications

Identifiants chimiques

Cell Imaging Multi-Mode Microplate Reader
Sony CMOS, 16-bit Gray Scale, Standard or WFOV
230 to 999 nm, 1 nm increments
Absorbance: 0 to 4 OD; Fluorescence Intensity: 7 decades; Luminesence: >6 decades
Fluorescence, phase contrast, brightfield, high contrast brightfield, color brightfield and phase contrast
2 Syringe Pumps
Xenon Flash; 100 mW 680 nm laser (Alpha Detection)
4-Zone incubation to 65°C with Condensation Control (±0.2°C at 37°C)
51.4 cm
44.5 cm
36.3 kg
41.6 cm
250 W max.
Linear, Orbital, Double Orbital
0 to 20%
Monochromator: ±2 nm; Dispenser: ±1 μL or 2%
BioStack and Third Party Automation Compatible; BioSpa 8 Automated Incubator Compatible
5 to 1000 μL, 1 μL increments
End point, kinetic, spectral scanning and well area scanning
1.25x, 2.5x (2.25x eff), 2.5x (2.75x eff), 4x, 10x, 20x, 40x, 60x
20.2 in.
17.5 in.
80 lb.
16.4 in.
CE and TUV marked. RoHS compliant.

Spécifications

Cell Imaging Multi-Mode Microplate Reader
Sony CMOS, 16-bit Gray Scale, Standard or WFOV
230 to 999 nm, 1 nm increments
Absorbance: 0 to 4 OD; Fluorescence Intensity: 7 decades; Luminesence: >6 decades
Fluorescence, phase contrast, brightfield, high contrast brightfield, color brightfield and phase contrast
Wide FOV camera; Fluorescence/brightfield/color brightfield imaging; Monochromator-based UV-Vis absorbance/fluorescence intensity/time resolved fluorescence (secondary)/luminescence
2 Syringe Pumps
End point, kinetic, spectral scanning and well area scanning
1.25x, 2.5x (2.25x eff), 2.5x (2.75x eff), 4x, 10x, 20x, 40x, 60x
4-Zone incubation to 65°C with Condensation Control (±0.2°C at 37°C)
51.4 cm
44.5 cm
36.3 kg
41.6 cm
250 W max.
Linear, Orbital, Double Orbital
0 to 20%
Monochromator: ±2 nm; Dispenser: ±1 μL or 2%
BioStack and Third Party Automation Compatible; BioSpa 8 Automated Incubator Compatible
5 to 1000 μL, 1 μL increments
CYT5MW
Monochromator: 6- to 384-well plates; Filters: 6-to 1536 well plates; Imaging: 6-to 1536 well plates; Take3 Plate compatible
Xenon Flash; 100 mW 680 nm laser (Alpha Detection)
Gen5 Microplate Reader and Imager Software included; Gen5 Image+ and Image Prime software available for full image analysis; Gen5 Secure for 21 CFR Part 11 compliance (option)
20.2 in.
17.5 in.
80 lb.
16.4 in.
CE and TUV marked. RoHS compliant.
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions