Learn More
Agilent BioTek Cytation 1 Cell Imaging Multi-Mode Reader

BioTek Cytation 1 cell imaging multi-mode reader combines fluorescence and high contrast brightfield imaging with conventional multi-mode detection. For Research Use Only. Not for use in diagnostic procedures.
Supplier: Agilent BioTek CYT1FASN
Description
BioTek Cytation 1 cell imaging multi-mode reader combines fluorescence and high contrast brightfield imaging with conventional multi-mode detection. This affordable imaging reader delivers performance and economy compared to other digital microscopy systems
The BioTek Cytation 1 design provides both quantitative phenotypic cellular data with well-based quantitative data. The multi-mode detection module has high sensitivity filter-based fluorescence and a monochromator for UV-Vis absorbance. Temperature control and shaking are standard. CO2/O2 control and reagent injectors are available. Gen5 software simplifies image capture and plate reading.
Identifiants chimiques
| Cell Imaging Multi-Mode Microplate Reader | |
| 0 to 20% | |
| 6-well to 384-well plates | |
| Xenon Flash | |
| Gen5 Microplate Reader and Imager Software | |
| 20 in. | |
| 17.5 in. | |
| 16.5 in. | |
| CE and TUV marked. RoHS compliant. |
| Linear, Orbital, Double Orbital | |
| 0 to 19% | |
| Endpoint, kinetic, area scanning, plate mode inject/read process and well mode inject/read process | |
| 0.3 μm/pixel at 20X | |
| Incubation to 45°C | |
| 50.8 cm | |
| 44.5 cm | |
| 41.9 cm |
Spécifications
| Cell Imaging Multi-Mode Microplate Reader | |
| 0 to 20% | |
| Monochromator absorbance; Filter/dichroic fluorescence; Luminescence/filtered luminescence | |
| 6-well to 384-well plates | |
| Xenon Flash | |
| Gen5 Microplate Reader and Imager Software | |
| 20 in. | |
| 17.5 in. | |
| 16.5 in. | |
| CE and TUV marked. RoHS compliant. |
| Linear, Orbital, Double Orbital | |
| 0 to 19% | |
| CYT1FA | |
| Endpoint, kinetic, area scanning, plate mode inject/read process and well mode inject/read process | |
| 0.3 μm/pixel at 20X | |
| Incubation to 45°C | |
| 50.8 cm | |
| 44.5 cm | |
| 41.9 cm |