missing translation for 'onlineSavingsMsg'
Learn More

Cystamine Dihydrochloride 97.0+%, TCI America™

Catalog No. C087525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C087525G 25 g
C0875100G 100 g
C0875500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C087525G Supplier TCI America Supplier No. C087525G

Chemical Identifiers

CAS 56-17-7
Molecular Formula C4H14Cl2N2S2
Molecular Weight (g/mol) 225.19
MDL Number MFCD00012905
InChI Key YUFRRMZSSPQMOS-UHFFFAOYSA-N
Synonym cystamine dihydrochloride, cystamin dihydrochloride, ethanamine, 2,2'-dithiobis-, dihydrochloride, usaf cb-34, 2,2'-disulfanediyldiethanamine dihydrochloride, 2-aminoethyl disulfide dihydrochloride, bis 2-aminoethyl disulfide dihydrochloride, cystamine 2hcl, 2,2'-dithiodi ethylammonium dichloride, decarboxycystine dihydrochloride
PubChem CID 5941
IUPAC Name 2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride
SMILES C(CSSCCN)N.Cl.Cl

Specifications

Melting Point 218°C
Color White
Quantity 25 g
Formula Weight 225.19
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Cystamine Dihydrochloride
RTECSNumber KR7260000
TSCA Yes